Formalin 10%, Q Path®, buffered
80 ml in a 150 ml plastic pot, 32
Fournisseur: VWR Collection
Formalin 10%, Q Path®, buffered
N° de catalogue VWRSFOR0151A800004
Fournisseur: VWR Collection
Numéro CAS: 50-00-0
Jump to:
Product Details & Documents
Formalin ready to use fixative for tissue samples. Pots and buckets can be used for storage.
Formaldehyde 4% is approximately equal to Formalin 10%
Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Density:1.013 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-001-8
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Appearance:Colourless liquid
- Test formaldehyde:Passes test
- Density (20/4):1,012 - 1,014
- pH (20°C):6,8 - 7,2
Specifications
- Pk:32
- Pk Info:80 ml in a 150 ml plastic pot
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



