Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mounting medium, PERTEX®

Mounting medium, PERTEX®

PERTEX®, mounting medium, 500 ml, 1
Catalog # LEIC811
Supplier:   LEICA MICROSYSTEMS
Mounting medium, PERTEX®
Catalog # LEIC811
Supplier:  LEICA MICROSYSTEMS
CAS Number:  
Mounting medium, PERTEX®

PERTEX®, mounting medium, 500 ml, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

PERTEX® is the original rapid drying medium for mounting and preserving of slide specimens. PERTEX® is proven not to affect the form, structure or colour of the mounted speci­men, even after years of storage. A thin coating of PERTEX® is enough for the cover glass to adhere to a microscope slide.

  • Xylene based medium, allows for immediate screening of slides
  • Low viscosity enable it to flow easily and prevent the formation of air bubbles
  • Neutral, colourless and has a refractive index close to that of glass at +20 °C

PERTEX® is soluble in xylene and most xylene substitutes, mounting from 99,9% ethanol is also possible.

Documents

Specifications

Specifications

  • Pk:
    1
  • Description:
    PERTEX®, mounting medium, 500 ml

Specifications