Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Formalin 10%, Q Path®, buffered

25 ml in a 60 ml plastic pot, 50
Catalog # VWRSFOR0061A670451
Health
Irritant
Supplier:   VWR Collection
Formalin 10%, Q Path®, buffered
Catalog # VWRSFOR0061A670451
Supplier:  VWR Collection
CAS Number:  50-00-0
Formalin 10%, Q Path®, buffered

25 ml in a 60 ml plastic pot, 50

CAS Number:  50-00-0
Price
Inquire For Price
Log in for availability
1 * 50 ST
Product Details & Documents

Formalin ready to use fixative for tissue samples. Pots and buckets can be used for storage.

Formaldehyde 4% is approximately equal to Formalin 10%

Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Density:1.013 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Specifications

Specification Test Results

  • Appearance:
    Colourless liquid
  • Test formaldehyde:
    Passes test
  • Density (20/4):
    1,012 - 1,014
  • pH (20°C):
    6,8 - 7,2

Specifications

  • Pk:
    50
  • Pk Info:
    25 ml in a 60 ml plastic pot

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.