Formalin 10%, Q Path®, buffered
5 ml in a 20 ml plastic pot, 102
Formalin ready to use fixative for tissue samples. Pots and buckets can be used for storage.
Formaldehyde 4% is approximately equal to Formalin 10%
: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
- :50-00-0
- :CH₂O
- :MFCD00003274
- :30.03 g/mol
- :1.013 g/cm³ (20 °C)
- :Ambient
- :200-001-8
- Appearance:Colourless liquid
- Test formaldehyde:Passes test
- Density (20/4):1,012 - 1,014
- pH (20°C):6,8 - 7,2
- Pk:102
- Pk Info:5 ml in a 20 ml plastic pot
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



