Formalin 10%, Q Path®, buffered
25 ml in a 60 ml plastic pot, 50
Supplier: VWR Collection
Formalin 10%, Q Path®, buffered
Catalog # VWRSFOR0061A691044
Supplier: VWR Collection
CAS Number: 50-00-0
Jump to:
Product Details & Documents
Formalin ready to use fixative for tissue samples. Pots and buckets can be used for storage.
Formaldehyde 4% is approximately equal to Formalin 10%
Synonyms: Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- Density:1.013 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-001-8
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Appearance:Colourless liquid
- Test formaldehyde:Passes test
- Density (20/4):1,012 - 1,014
- pH (20°C):6,8 - 7,2
Specifications
- Pk:50
- Pk Info:25 ml in a 60 ml plastic pot
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



