Order Entry
Belgium
Orders LinkContactUsLinkComponent
Formalin 10%, Q Path®, buffered

Formalin 10%, Q Path®, buffered

25 ml in a 60 ml plastic pot, 50
N° de catalogue VWRSFOR0061AB11045
Health
Irritant
Fournisseur:   VWR Collection
Formalin 10%, Q Path®, buffered
N° de catalogue VWRSFOR0061AB11045
Fournisseur:  VWR Collection
Numéro CAS:  50-00-0
Formalin 10%, Q Path®, buffered

25 ml in a 60 ml plastic pot, 50

Numéro CAS:  50-00-0
Prix
Inquire For Price
Log in for availability
1 * 50 ST
Product Details & Documents

Formalin ready to use fixative for tissue samples. Pots and buckets can be used for storage.

Formaldehyde 4% is approximately equal to Formalin 10%

Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Density:1.013 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Specifications

Specification Test Results

  • Appearance:
    Colourless liquid
  • Test formaldehyde:
    Passes test
  • Density (20/4):
    1,012 - 1,014
  • pH (20°C):
    6,8 - 7,2

Specifications

  • Pk:
    50
  • Pk Info:
    25 ml in a 60 ml plastic pot

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.