Orders LinkContactUsLinkComponent
Test charts
Test charts
Test charts with optical brightner, Black/white chequered, A3, 297×420 mm
Catalogus # SIMEVF2354
Leverancier:   Industrial Physics
Test charts
Catalogus # SIMEVF2354
Leverancier:  Industrial Physics
Test charts
Test charts
Test charts with optical brightner, Black/white chequered, A3, 297×420 mm
Price
282,00 €
Log in voor beschikbaarheid
1 * 250 ST
Product Details & Documents
Test charts are used for testing physical properties of coating, lacquers, and inks.
  • Dimensions from A6 up to A4
  • Film laminated for an excellent solvent, chemical resistance, and film spread
  • Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Bestelinformatie: A certificate is available for charts with an optical brightener.
Specificaties
  • Format:
    A3
  • Colour:
    Black/white chequered
  • Description:
    Test charts with optical brightner
  • Seq:
    11
  • W×H:
    297×420 mm