Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH Buffer solutions, CX-1

pH Buffer solutions, CX-1

pH gradient buffer, CX-1, buffer B, 10,2, 250 mL
Catalog # 555-9511
pH Buffer solutions, CX-1
Catalog # 555-9511
CAS Number:  
pH Buffer solutions, CX-1
pH Buffer solutions, CX-1

pH gradient buffer, CX-1, buffer B, 10,2, 250 mL

CAS Number:  
Price
348,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Delivers robust reproducible gradients with the Thermo Scientific™ pH buffer platform that is applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    pH gradient buffer, CX-1, buffer B
  • pH value:
    10,2

Specifications