Order Entry
Belgium
Orders LinkContactUsLinkComponent
pH Buffer solutions, CX-1

pH Buffer solutions, CX-1

pH gradient buffer, CX-1, buffer A, 5,6, 125 mL
Catalog # 555-9504
pH Buffer solutions, CX-1
Catalog # 555-9504
CAS Number:  
pH Buffer solutions, CX-1
pH Buffer solutions, CX-1

pH gradient buffer, CX-1, buffer A, 5,6, 125 mL

CAS Number:  
Price
217,00 €
Log in for availability
1 * 125 mL
Product Details & Documents

Delivers robust reproducible gradients with the Thermo Scientific™ pH buffer platform that is applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.

Specifications

Specifications

  • Pk:
    125 mL
  • Description:
    pH gradient buffer, CX-1, buffer A
  • pH value:
    5,6

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.