Orders LinkContactUsLinkComponent
Extendable samplers, TeleSwab
Telescopic swabbing tool with extendable pole, swabbing template, template holder and angle adapter
Catalog # 300-0421
Supplier:   Sampling Systems
Extendable samplers, TeleSwab
Catalog # 300-0421
Supplier:  Sampling Systems
Extendable samplers, TeleSwab
Extendable samplers, TeleSwab
Telescopic swabbing tool with extendable pole, swabbing template, template holder and angle adapter
Price
3 920,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The TeleSwab has an extendable handle, which allows to take swabs from high and difficult to reach places.
  • Environmental swabbing for use with large items of processing equipment
  • Extendable handle to the correct length for easy swabbing
  • Adapter allowing the swabs to be angled to the contours of the sampled item
 : Swabs are not included with these kits.
Specifications
  • Description:
    Telescopic swabbing tool with extendable pole, swabbing template, template holder and angle adapter