Extendable samplers, TeleSwab
Telescopic swabbing tool with extendable pole, swab holder and angle adapter
Catalog # 300-0420
Supplier: Sampling Systems
Jump to:
Product Details & Documents
The TeleSwab has an extendable handle, which allows to take swabs from high and difficult to reach places.
- Environmental swabbing for use with large items of processing equipment
- Extendable handle to the correct length for easy swabbing
- Adapter allowing the swabs to be angled to the contours of the sampled item
Ordering information: Swabs are not included with these kits.
Specifications
- Description:Telescopic swabbing tool with extendable pole, swab holder and angle adapter