Orders LinkContactUsLinkComponent
Detectable plasters
Detectable plasters
Detectable plaster strips, blue, refill set for dispenser, 40 pieces, 72×25 mm
Catalog # SOEH1009984
Supplier:   SOEHNGEN
Detectable plasters
Catalog # SOEH1009984
Supplier:  SOEHNGEN
Detectable plasters
Detectable plaster strips, blue, refill set for dispenser, 40 pieces, 72×25 mm
Price
13,90 €
Log in for availability
1 * 1 KIT
Product Details & Documents
These blue, detectable plasters are specially designed for use in the food industry. Dispenser containing aluminium strips with an integrated metal surface that can be detected via a metal detector.
  • Plasters are detectable by most metal detectors used within the food industry
  • Breathable and skin-friendly
  • High quality hypoallergenic plasters
Specifications
  • Description:
    Detectable plaster strips, blue, refill set for dispenser, 40 pieces
  • L×W:
    72×25 mm
  • Seq:
    03