Ultra Yield™ Flasks
Product Details & Documents
Thomson's Ultra Yield™ flasks are so named because these flasks actually enhance the growth of E.coli as well as other microbial cells.
The AirOtop® Seals are designed as a single-use, disposable seal. Enhanced AirOtop® Seals offer simple, disposable, sterile high air exchange. Seals fit all shake flasks. The Enhanced AirOtop® Seal's properties include a 0,2 µm sterile barrier with a long-term resealable membrane for use with Ultra Yield™ and other brand flasks. Testing has been conducted on multiple customer samples with great results on up to seven days of cellular growth. Testing included Protista (Algae), E.coli, and other microbes resulting in improved growth.
The patented design of the flasks renders them to an improved microbial cell yield. Flasks are available in standardized sizes of 125, 250 L, 500 and 2500 ml. The flasks can be closed snug using the AirOTop seals. These seals are designed to fit on the flask-tops. Aeration caps are also available for long-term use on flasks for applications such as CHO and other suspension cell growth procedures.
Documents
Certificates
Search by Lot NumberCell Culture Solutions Ultra Yield System (Microbial and E. coli Cell Growth).pdf
Additional Product Details
Specifications
- Capacity:250 ml
- Description:Ultra yield flask
- Seq:4
Accessories
Product Information
- Catalog #AvailabilityPrice
- AirOtop enhanced seal for 2,5 L ultra yield flaskAirOtop enhanced seal for 2,5 L ultra yield flaskTHMS899425Log in for availability1 * 100 ST - 340,00 €
Product Information
- Catalog #AvailabilityPrice
- AirOtop enhanced seal for 125 ml ultra yield flaskAirOtop enhanced seal for 125 ml ultra yield flaskTHMS899421_ERRLog in for availability1 * 100 ST
Product Information
- Catalog #AvailabilityPrice
- AirOtop enhanced seal for 500 ml ultra yield flaskAirOtop enhanced seal for 500 ml ultra yield flaskTHMS899424Log in for availability1 * 100 ST - 301,00 €
Product Information
- Catalog #AvailabilityPrice
- AirOtop enhanced seal for 250 ml ultra yield flaskAirOtop enhanced seal for 250 ml ultra yield flaskTHMS899423Log in for availability1 * 100 ST - 301,00 €
Product Information
- Catalog #AvailabilityPrice
- AirOtop enhanced seal for 125 ml ultra yield flask



