Order Entry
Belgium
Orders LinkContactUsLinkComponent
ORP test solution

ORP test solution

ORP test solution, 600 mV, Plastic bottle, 500 mL
Catalog # HACH25M2A1002-115
Supplier:   Hach
ORP test solution
Catalog # HACH25M2A1002-115
Supplier:  Hach
CAS Number:  
ORP test solution
ORP test solution

ORP test solution, 600 mV, Plastic bottle, 500 mL

CAS Number:  
Price
74,80 €
Log in for availability
1 * 500 mL
Product Details & Documents

ORP (Oxidation Reduction Potential) reference solution used for ORP sensor calibration.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Description:
    ORP test solution, 600 mV

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.