Volumetric Flasks, Class A
20 ml, 14/23, 0,10 ± ml
Catalog # WITG3.664.00214
Supplier: witeg Labortechnik
Jump to:
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
: Tolerance according to DIN EN ISO 1042
Specifications
- NS:14/23
- Capacity:20 ml
- Seq:16
- Tolerance:0,10 ± ml