ContactUsLinkComponent
10mg C/L Carbon Verification Set consisting of 1 x 40ml vial each of calibration blank (RTOCW), 10mg/L (RTOCS05) TOC C as Sucrose an d 10mg/L (RTICN06) TIC C as Na2CO3 1 * 40 mL
Catalog # REGERC120018
Supplier:  REAGECON
10mg C/L Carbon Verification Set consisting of 1 x 40ml vial each of calibration blank (RTOCW), 10mg/L (RTOCS05) TOC C as Sucrose an d 10mg/L (RTICN06) TIC C as Na2CO3 1 * 40 mL
Catalog # REGERC120018
Supplier:  REAGECON
Frequently Bought Together