ContactUsLinkComponent
5mg C/L Carbon Verification Set consisting of 1 x 40ml vial each of calibration blank (RTOCW), 5mg/L (RTOCS04) TOC C as Sucrose and 5 mg/L (RTICN05) TIC C as Na2CO3 1 * 40 mL
Catalog # REGERC120006
Supplier:  REAGECON
5mg C/L Carbon Verification Set consisting of 1 x 40ml vial each of calibration blank (RTOCW), 5mg/L (RTOCS04) TOC C as Sucrose and 5 mg/L (RTICN05) TIC C as Na2CO3 1 * 40 mL
Catalog # REGERC120006
Supplier:  REAGECON
Frequently Bought Together