Order Entry
Belgium
Orders LinkContactUsLinkComponent
Osmolality standards

Osmolality standards

Osmolality standard, 200 mOsm/kg H₂O, Ampoule, 12×5 ml, 1 KIT
Catalog # REGERE-OSM-200
Supplier:   REAGECON
Osmolality standards
Catalog # REGERE-OSM-200
Supplier:  REAGECON
CAS Number:  
Osmolality standards

Osmolality standard, 200 mOsm/kg H₂O, Ampoule, 12×5 ml, 1 KIT

CAS Number:  
Price
204,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Osmolality standards provide ease of use when calibrating all types of osmometers. All Osmolality standards are manufactured using high purity raw materials. These products are prepared gravimetrically on a weight/weight basis. Both solute (salts) and solvent (water) are weighed on a balance calibrated by using OIML traceable weights.

  • Ready to use and presented in convenient ampoules
  • Extended shelf life and has low uncertainty of measurement
  • Consistency of product-Independent, traceable
  • Consist of 12 ampoules of 5 ml

The Osmolality of the standard is verified using a high performance calibrated, temperature controlled osmometer. The calibration of this instrument is completed using high purity certified Osmolality standards.

Certifications: ISO17025 accreditation for calibration of non automatic weighing machines (INAB ref: 265C). The resulting Balance Certificate of Calibration is issued in accordance with the requirements of ISO/IEC 17025.

Documents

Specifications

Specifications

  • Pack type:
    Ampoule
  • Pk:
    1 KIT
  • Description:
    Osmolality standard
  • Pk Info:
    12×5 ml
  • Osmolality:
    200 mOsm/kg H₂O

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.