Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Phosphorus standard solution

Phosphorus standard solution, 100 ml (NIST), 1000 mg/l as P, 100 mL
Catalog # HACH2321142
Supplier:   Hach
Phosphorus standard solution
Catalog # HACH2321142
Supplier:  Hach
CAS Number:  
Phosphorus standard solution
Phosphorus standard solution

Phosphorus standard solution, 100 ml (NIST), 1000 mg/l as P, 100 mL

CAS Number:  
Price
21,90 €
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solution for accuracy checks in phosphorus determinations.

Specifications

Specifications

  • Concentration:
    1000 mg/l as P
  • Pk:
    100 mL
  • Description:
    Phosphorus standard solution, 100 ml (NIST)

Specifications

Aanbevelingen

Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.

Anders ontvangt u algemene aanbevelingen.