Order Entry
Belgium
Orders LinkContactUsLinkComponent
Electrolyte solutions, aqueous

Electrolyte solutions, aqueous

Electrolyte solution L3004 cert - KCl 3.0 mol/l, sterilized, 250 ml in DURAN® glass bottle, 250 mL
Catalog # SCOI285138440
Supplier:   SI Analytics
Electrolyte solutions, aqueous
Catalog # SCOI285138440
Supplier:  SI Analytics
CAS Number:  
Electrolyte solutions, aqueous
Electrolyte solutions, aqueous

Electrolyte solution L3004 cert - KCl 3.0 mol/l, sterilized, 250 ml in DURAN® glass bottle, 250 mL

CAS Number:  
Price
55,10 €
Log in for availability
1 * 250 mL
Product Details & Documents

Electrolyte aqueous solutions are used for reference electrodes, as electrolyte bridges and for storage.

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    Electrolyte solution L3004 cert - KCl 3.0 mol/l, sterilized, 250 ml in DURAN® glass bottle

Specifications