Order Entry
Belgium
Orders LinkContactUsLinkComponent
Closures for test tubes and flasks

Closures for test tubes and flasks

Identi-plug foam plugs, Ø 6 - 13 mm
Catalog # JAECL800-A
Supplier:   JAECE INDUSTRIES
Closures for test tubes and flasks
Catalog # JAECL800-A
Supplier:  JAECE INDUSTRIES
Closures for test tubes and flasks
Closures for test tubes and flasks

Identi-plug foam plugs, Ø 6 - 13 mm

Price
174,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

Identy-plug.

Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.

Cotton cheesecloth.

100% pure cotton, very absorbent and low linting; 28×24 thread count.

Bleached cotton balls.

Available large or medium.

Specifications
  • Description:
    Identi-plug foam plugs, Ø 6 - 13 mm
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.