ContactUsLinkComponent
Shelf for star-desiccator, pmma, incl. 2 rails made of polyamide, suitable for all sicco-star-desiccators (except model horizontal) 1 * 1 items
Catalog # BOHLV1860-68
Supplier:  Bohlender
undefined
Shelf for star-desiccator, pmma, incl. 2 rails made of polyamide, suitable for all sicco-star-desiccators (except model horizontal) 1 * 1 items
Catalog # BOHLV1860-68
Supplier:  Bohlender
Frequently Bought Together