Sodium nitrate, extra pure
Plastic bottle, 1 kg
Leverancier: Thermo Fisher Scientific
Sodium nitrate, extra pure
Catalogus # ACRO436090010
Leverancier: Thermo Fisher Scientific
CAS-nummer: 7631-99-4
Ga naar:
Product Details & Documents
- CAS-nummer:7631-99-4
- Formule:NaNO₃
- MDL-nummer:MFCD00011119
- Molecular Weight:84.99 g/mol
- Kokend Pt:380 °C (760 mmHg)
- Smelten Pt:306 °C
- Dichtheid:2,26 g/cm³ (20 °C)
- Temperatuur bij opslag:Ambient
- UN:1498
- ADR:5.1,III
Documents
ACRO43609
SDS
Specificaties
Specificatie testresultaten
- Appearance:Colorless to white Crystalline powder or crystals
- Loss on drying:≤2 % (105°C)
- pH:5.5 to 8.3 (5 % in water)
- Titration after Ion exchange:≥99.0 %
Specificaties
- Pack type:Plastic bottle
- Pk:1 kg



