Order Entry
Belgium
Orders LinkContactUsLinkComponent
Media in Bottles

Media in Bottles

Nacl Peptone buffer + 0.1% Tween 80, —, 1000 ml, Without cap, 1000 ml, 6
Catalog # HEIP1463810006
Supplier:   MERCK LIFE SCIENCE
Media in Bottles
Catalog # HEIP1463810006
Supplier:  MERCK LIFE SCIENCE
CAS Number:  
Media in Bottles
Media in Bottles

Nacl Peptone buffer + 0.1% Tween 80, —, 1000 ml, Without cap, 1000 ml, 6

CAS Number:  
Price
114,00 €
Log in for availability
1 * 6 ST
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    6
  • Standards:
  • Description:
    Nacl Peptone buffer + 0.1% Tween 80
  • Size:
    1000 ml
  • Closure type:
    Without cap
  • Filling:
    1000 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.