Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Media in Bottles

Nacl Peptone buffer + 0.1% Tween 80, —, 90 ml, Without cap, 125 ml, 6
Catalog # HEIP1463710006
Supplier:   MERCK LIFE SCIENCE
Media in Bottles
Catalog # HEIP1463710006
Supplier:  MERCK LIFE SCIENCE
CAS Number:  
Media in Bottles
Media in Bottles

Nacl Peptone buffer + 0.1% Tween 80, —, 90 ml, Without cap, 125 ml, 6

CAS Number:  
Price
47,20 €
Log in for availability
1 * 6 ST
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    6
  • Standards:
  • Description:
    Nacl Peptone buffer + 0.1% Tween 80
  • Size:
    125 ml
  • Closure type:
    Without cap
  • Filling:
    90 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.