Order Entry
Belgium
Orders LinkContactUsLinkComponent
Bouffant caps

Bouffant caps

Green, 520 mm
Catalog # 113-9211
Supplier:   VWR Collection
Bouffant caps
Catalog # 113-9211
Supplier:  VWR Collection
Bouffant caps
Bouffant caps

Green, 520 mm

Price
Inquire For Price
Log in for availability
1 * 1.000 ST
Product Details & Documents

These disposable caps made of PP are ideal for use in non critical environments with high hygiene standards.

  • Lightweight fabric
  • Latex-free elastic band
  • Available in different colours and sizes

Weight: 10 to 12 g/m²

Packaging: 100 pieces per double polybag; 1000 pieces per carton.

Specifications
  • Ø:
    520 mm
  • Colour:
    Green
  • Cat. No.:
    113-9211
  • Garment application:
    General Purpose
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.