Bouffant caps
White, 520 mm
113-9209
These disposable caps made of PP are ideal for use in non critical environments with high hygiene standards.
- Lightweight fabric
- Latex-free elastic band
- Available in different colours and sizes
Weight: 10 to 12 g/m²
: 100 pieces per double polybag; 1000 pieces per carton.
- Ø:520 mm
- Colour:White
- Cat. No.:113-9209
- Garment application:General Purpose



