Orders LinkContactUsLinkComponent
Stirrer Guides with Anti-clockwise Thread
Stirrer Guides with Anti-clockwise Thread
Stirrer guide, dynamic double seal, 29/32, 16 mm
Catalog # LENZ06306204
Supplier:   Lenz Laborglas
Stirrer Guides with Anti-clockwise Thread
Catalog # LENZ06306204
Supplier:  Lenz Laborglas
Stirrer Guides with Anti-clockwise Thread
Stirrer Guides with Anti-clockwise Thread
Stirrer guide, dynamic double seal, 29/32, 16 mm
Price
390,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, PFTE/fibre glass pressure screw. The fluoroplastic bearing is in a borosilicate glass housing, and has adjustable PTFE/FPM seal. All parts are interchangeable.
Spécifications
  • NS:
    29/32
  • Description:
    Stirrer guide, dynamic double seal
  • Seq:
    03
  • Shaft Ø:
    16 mm