Order Entry
Belgium
Orders LinkContactUsLinkComponent
Gas sampling balloons

Gas sampling balloons

Gas sampling balloon, 250 mm, 8,0 L
Catalog # KART1235
Supplier:   KARTELL
Gas sampling balloons
Catalog # KART1235
Supplier:  KARTELL
Gas sampling balloons
Gas sampling balloons

Gas sampling balloon, 250 mm, 8,0 L

Price
80,70 €
Log in for availability
1 * 1 ST
Product Details & Documents

Gas collection bags made from latex.

Designed for general use in laboratories.

Ordering information: Available in different volumes and diameters.

Documents

Specifications
  • Ø:
    250 mm
  • Volume:
    8,0 L
  • Description:
    Gas sampling balloon