ContactUsLinkComponent
Loading Trolley Large for RSC/RSV Models - For Priorclave 450 liter and 700 liter front loaded models with rectangular chamber 1 * 1 items
Catalog # PRIOPC/TLY/RSL
Supplier:  Priorclave
Loading Trolley Large for RSC/RSV Models - For Priorclave 450 liter and 700 liter front loaded models with rectangular chamber 1 * 1 items
Catalog # PRIOPC/TLY/RSL
Supplier:  Priorclave
Frequently Bought Together