ContactUsLinkComponent
Slide with dual slit, this generates a phenomenon of interference between the diffraction figures of each slit, this experiment is c alled young slits experiment, the slide contains two slots of 50mm , with a gap of 0.1mm. 1 * 1 items
Catalog # DIDAPOM052820
Supplier:  DIDALAB
Slide with dual slit, this generates a phenomenon of interference between the diffraction figures of each slit, this experiment is c alled young slits experiment, the slide contains two slots of 50mm , with a gap of 0.1mm. 1 * 1 items
Catalog # DIDAPOM052820
Supplier:  DIDALAB
Frequently Bought Together