ContactUsLinkComponent
Oscilloscope, digital, bandwidth 500MHz, 2channels, 25k points memory for each channel, visual persistence oscilloscope, 8 inches sc reen, 800x600 high resolution TFT LCD display, built-in three inpu t impedance selection 50o aohms. 1 * 1 items
Catalog # 761-1003
Supplier:  DIDALAB
undefined
Oscilloscope, digital, bandwidth 500MHz, 2channels, 25k points memory for each channel, visual persistence oscilloscope, 8 inches sc reen, 800x600 high resolution TFT LCD display, built-in three inpu t impedance selection 50o aohms. 1 * 1 items
Catalog # 761-1003
Supplier:  DIDALAB
Frequently Bought Together