Order Entry
Belgium
Orders LinkContactUsLinkComponent
Microscope objective lens mounts

Microscope objective lens mounts

Lens holder with angular adjustment
Catalog # DIDAPOD060560
Supplier:   DIDALAB
Microscope objective lens mounts
Catalog # DIDAPOD060560
Supplier:  DIDALAB
Microscope objective lens mounts
Microscope objective lens mounts

Lens holder with angular adjustment

Price
91,88 €
Log in for availability
1 * 1 ST
Product Details & Documents

The objective holder allows precise positioning of a microscope objective according to the vertical and the horizontal.

  • Both axes 2 mm each
  • Holder is mounted on a 10 mm diameter rod
Specifications
  • Description:
    Lens holder with angular adjustment
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.