ContactUsLinkComponent
Sodium spectral lamp, power supply + low pressure, 18W, adapted ventilation, 2P+E power outlet with fuse, no risk of burning, econom icalm, can be used on the lamp base or on optical bench, UK. 1 * 1 items
Catalog # DIDAPOD010058_UK
Supplier:  DIDALAB
Sodium spectral lamp, power supply + low pressure, 18W, adapted ventilation, 2P+E power outlet with fuse, no risk of burning, econom icalm, can be used on the lamp base or on optical bench, UK. 1 * 1 items
Catalog # DIDAPOD010058_UK
Supplier:  DIDALAB
Frequently Bought Together