ContactUsLinkComponent
Oscilloscope, digital, bandwidth 250MHz, 2 channels, 25k points memory for each channel, visual persistence oscilloscope, 8inches sc reen, 800x600 high resolution TFT LCD display, built-in 3 input impedance selection, 50ohms/75ohms/1m 1 * 1 items
Catalog # 761-1001
Supplier:  DIDALAB
undefined
Oscilloscope, digital, bandwidth 250MHz, 2 channels, 25k points memory for each channel, visual persistence oscilloscope, 8inches sc reen, 800x600 high resolution TFT LCD display, built-in 3 input impedance selection, 50ohms/75ohms/1m 1 * 1 items
Catalog # 761-1001
Supplier:  DIDALAB
Frequently Bought Together