ContactUsLinkComponent
Slit, adjustable, with symmetric opening, stainless steel lips, opening up to 4mm over a height of 15mm, aperture adjustment by one hundredth of a micrometer screw, reading the micrometer corresponds to the actual width of the slot. 1 * 1 items
Catalog # DIDAPOD070310
Supplier:  DIDALAB
Slit, adjustable, with symmetric opening, stainless steel lips, opening up to 4mm over a height of 15mm, aperture adjustment by one hundredth of a micrometer screw, reading the micrometer corresponds to the actual width of the slot. 1 * 1 items
Catalog # DIDAPOD070310
Supplier:  DIDALAB
Frequently Bought Together