Optical components for optical bench
Direct vision prism with internal Amici prism
Catalog # DIDAPOD068282
Supplier: DIDALAB
Jump to:
Product Details & Documents
Specifications
- Description:Direct vision prism with internal Amici prism
Accessories
Product Information
- Catalog #AvailabilityPrice
- Permanent frame, Ø 40 mm
Product Information
- Catalog #AvailabilityPrice
- Simple lens holder, Ø 40 mm
Product Information
- Catalog #AvailabilityPrice
- Lens holder for fragile components, Ø 80 mmLens holder for fragile components, Ø 80 mmDIDAPOD010080Log in for availability1 * 1 ST - 75,25 €
Product Information
- Catalog #AvailabilityPrice
- Lens holder, Ø 40/42 mm
Product Information
- Catalog #AvailabilityPrice
- Lens holder, Ø 80 mm
Product Information
- Catalog #AvailabilityPrice
- Stand, half moon, for optical components positioned remote from the Optical bench, height 210 mmStand, half moon, for optical components positioned remote from the Optical bench, height 210 mmDIDAPOD002192Log in for availability1 * 1 ST - 126,00 €
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



