ContactUsLinkComponent
Gratings transmission, 200 lines/mm, these accuracy gratings have a great uniformity of lines on the whole working area, the working area (45x31mm) is protected by a glass plate, compatible with our slide-holders. 1 * 1 items
Catalog # DIDAPOD061990
Supplier:  DIDALAB
Gratings transmission, 200 lines/mm, these accuracy gratings have a great uniformity of lines on the whole working area, the working area (45x31mm) is protected by a glass plate, compatible with our slide-holders. 1 * 1 items
Catalog # DIDAPOD061990
Supplier:  DIDALAB
Frequently Bought Together