ContactUsLinkComponent
Prism crown, n=1.52, this prism has an equilateral stand, 40mm side, 48mm height, both working sides are highly polished as the othe r sides are dull. 1 * 1 items
Catalog # DIDAPOD068030
Supplier:  DIDALAB
Prism crown, n=1.52, this prism has an equilateral stand, 40mm side, 48mm height, both working sides are highly polished as the othe r sides are dull. 1 * 1 items
Catalog # DIDAPOD068030
Supplier:  DIDALAB
Frequently Bought Together