ContactUsLinkComponent
Prism flint, n=1.75, this prism has an equilateral stand, 40mm side, 48mm height, both working sides are highly polished as the othe r sides are dull. 1 * 1 items
Catalog # DIDAPOD068020
Supplier:  DIDALAB
Prism flint, n=1.75, this prism has an equilateral stand, 40mm side, 48mm height, both working sides are highly polished as the othe r sides are dull. 1 * 1 items
Catalog # DIDAPOD068020
Supplier:  DIDALAB
Frequently Bought Together