Dip slides
Dipslide Dermatest, 10
Catalog # LIOF50021
Supplier: LIOFILCHEM
Jump to:
Product Details & Documents
Flexible contact slide for microbiology screening of surfaces.
- ISO 18593
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pk:10
- Description:Dipslide Dermatest