Orders LinkContactUsLinkComponent
Flat mops
Flat mops
PolyTex™ mop heads, 45,72 cm
Catalog # MICRMZFM7-18
Supplier:   MICRONOVA
Flat mops
Catalog # MICRMZFM7-18
Supplier:  MICRONOVA
Flat mops
Flat mops
PolyTex™ mop heads, 45,72 cm
Price
34,80 €
Log in for availability
1 * 1 ST
Product Details & Documents
These brick-style, flat sponge mop heads are excellent for applying disinfectant in pharmaceutical production or aseptic processing applications.
  • Easily clean large surfaces
  • Heavy-duty construction offers additional resistance to textured floors and aggressive cleaning methods
  • Fully padded mop heads can be laundered and reused several times
Ideal for cleaning large surface areas as well as reaching shelving, doorjambs, and corners.
Mop heads feature heavy-duty construction with PolyTex™ textured polyester material, PolyTex™ material combined with MegaTex™ textured non-woven polyamide fabric strip, or PolyTex™ material combined with microfibre fabric strip. PolyTex™/MegaTex™ mop heads offer additional resistance to textured floors and more aggressive action for spills and heavy soils and these heads allow faster pick up of moisture in standard clean/disinfect applications such as hospitals and animal labs.
Ordering information: All components must be purchased separately. An electropolished stainless steel quick-disconnect adapter is required to attach mop head to an electropolished stainless steel extension handle.
Specifications
  • Description:
    PolyTex™ mop heads
  • Length:
    45,72 cm
  • Seq:
    05