Order Entry
Belgium
Orders LinkContactUsLinkComponent
Multi-element standards for ICP

Multi-element standards for ICP

Multi-element standards for 33 elements, 5% HNO₃, 100 mL
Catalog # ACSDM8A96.K1.5N.L1
Supplier:   CPACHEM
Multi-element standards for ICP
Catalog # ACSDM8A96.K1.5N.L1
Supplier:  CPACHEM
CAS Number:  
Multi-element standards for ICP

Multi-element standards for 33 elements, 5% HNO₃, 100 mL

CAS Number:  
Price
380,00 €
Log in for availability
1 * 100 mL
Product Details & Documents

ICP standards used for Al, Ag, As, B, Ba, Be, Bi, Ca, Cd, Cs, Co, Cr, Cu, Fe, In, K, Li, Mg, Mn, Mo, Na, Ni, Nb, Pb, Rb, Sb, Se, Sr, Ti, Tl, V, U, Zn.

Specifications

Specifications

  • Matrix:
    5% HNO₃
  • Pk:
    100 mL
  • Description:
    Multi-element standards for 33 elements

Specifications