Orders LinkContactUsLinkComponent
Jars with screw cap
with polypropylene screw cap, individually wrapped, labelled, irradiated, 150 ml
  216-2052
 :   Anicrin
Jars with screw cap
  216-2052
 :  Anicrin
Jars with screw cap
Jars with screw cap
with polypropylene screw cap, individually wrapped, labelled, irradiated, 150 ml
111,00 €
1 * 250 ST
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
  • Twist cap for quick opening and closing
  • Very good temperature resistance
  • Very good chemical resistance
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
  • Type:
    with polypropylene screw cap, individually wrapped, labelled, irradiated
  • Capacity:
    150 ml
  • Seq:
    08
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.