Human AHSA1 full-length ORF ( AAH00321, 1 A.A. - 338 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00010598-P012
Fournisseur: Abnova
Human AHSA1 full-length ORF ( AAH00321, 1 A.A. - 338 A.A.) Recombinant protein with GST-TAG AT N-terminal. 1 * 25 µG
Catalog # ABNOH00010598-P012
Fournisseur: Abnova
Frequently Bought Together
VWR® Facial Tissues
Fournisseur: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Fournisseur: VWR Chemicals
VWR®, Nitrile Gloves
Fournisseur: VWR Collection
pH indicator strips, MQuant®, Supelco®
Fournisseur: Merck
Sealing film for manual application, Parafilm® M
Fournisseur: HEATHROW
Acetone ≥99%, TECHNICAL
Fournisseur: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Fournisseur: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Fournisseur: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Fournisseur: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Fournisseur: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Fournisseur: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Fournisseur: VWR Collection