Storage Boxes for Vials
10×10, Green, 140×153×55 mm
N° de catalogue NEOL2-1937
Fournisseur: NEOLAB
Jump to:
Product Details & Documents
Storage boxes, made of PP, coloured, for HPLC vials, cryotubes or 1,5 and 2,0 ml microcentrifuge tubes with the height of 46 mm and up to 12 mm diameter.
- Stackable
- Transparent, tight sealing and hinged lid
- Also usable as cryobox
- Autoclavable
Specifications
- Compartments:10×10
- W×D×H ext.:140×153×55 mm
- Colour:Green
- Seq:03



