Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Sample dippers

Dip sampler for liquid samples, Transparent, PP, 300 cm, 22 mm
  BURK5315-0300
 :   BURKLE
Sample dippers
  BURK5315-0300
 :  BURKLE
Sample dippers
Sample dippers

Dip sampler for liquid samples, Transparent, PP, 300 cm, 22 mm

 
202,00 €
   
1 * 1 ST
 

Plunging siphons for the simple removal of liquid samples.

  • Made from transparent PP
  • All-layer and point sampling
  • Upper end with reflux valve, lower end with ball valve
 
  • Ø:
    22 mm
  • Colour:
    Transparent
  • Description:
    Dip sampler for liquid samples
  • Length:
    300 cm
  • Material:
    PP
  • Seq:
    04