Orders LinkContactUsLinkComponent
Tanks, rectangular, LDPE, LaboPlast®
Tanks, rectangular, LDPE, LaboPlast®
45 L, Seamless containers, rectangular shaped, - mm
Catalog # BURK3440-0455
Supplier:   BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Catalog # BURK3440-0455
Supplier:  BURKLE
Tanks, rectangular, LDPE, LaboPlast®
Tanks, rectangular, LDPE, LaboPlast®
45 L, Seamless containers, rectangular shaped, - mm
Price
340,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
These seamless rectangular tanks feature optimum ratio of wall thickness to filling load.
  • Linear LDPE, transparent
  • Rupture-proof
Specifications
  • Capacity:
    45 L
  • Description:
    Seamless containers, rectangular shaped
  • Seq:
    03
  • W×D×H:
    - mm
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Container thread ¾" for cylindrical containers
    Container thread ¾" for cylindrical containers
    BURK3440-0034
    Log in for availability
    1 * 1 ST - 213,00 €