Order Entry
Belgium
Orders LinkContactUsLinkComponent
Flow meters

Flow meters

Flow meter for fuel oil, diesel, hydrochloric acid, sodium hydroxide with connector thread G 3/4", 5 - 90 l/min, ±1 %, PP, Hastelloy C4, FKM, max.4 bar
Catalog # BURK5700-0111
Supplier:   BURKLE
Flow meters
Catalog # BURK5700-0111
Supplier:  BURKLE
Flow meters
Flow meters

Flow meter for fuel oil, diesel, hydrochloric acid, sodium hydroxide with connector thread G 3/4", 5 - 90 l/min, ±1 %, PP, Hastelloy C4, FKM, max.4 bar

Price
1 340,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These flow meters are compact and fully electronic for electric drum and container pumps, also used for low viscosity media.

  • Measurement by the turbine wheel principle
  • Can be calibrated depending on the medium
  • Different versions for many different applications
  • Automatic switch on when flow is recognized
  • Four resettable part ranges and one fixed total range
Specifications
  • Flow rate:
    5 - 90 l/min
  • Description:
    Flow meter for fuel oil, diesel, hydrochloric acid, sodium hydroxide with connector thread G 3/4"
  • Material:
    PP, Hastelloy C4, FKM
  • Seq:
    01
  • Accuracy:
    ±1 %
  • Pressure:
    max.4 bar
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.