Orders LinkContactUsLinkComponent
Detectable plasters
Detectable plasters
Detectable finger joint plasters, blue, refill set for dispenser, 20 plasters, - mm, 160×57×122 mm
Catalog # SOEH1009986
Supplier:   SOEHNGEN
Detectable plasters
Catalog # SOEH1009986
Supplier:  SOEHNGEN
Detectable plasters
Detectable finger joint plasters, blue, refill set for dispenser, 20 plasters, - mm, 160×57×122 mm
Price
Inquire For Price
Log in for availability
1 * 20 ST
Product Details & Documents
These blue, detectable plasters are specially designed for use in the food industry. Dispenser containing aluminium strips with an integrated metal surface that can be detected via a metal detector.
  • Plasters are detectable by most metal detectors used within the food industry
  • Breathable and skin-friendly
  • High quality hypoallergenic plasters
Specifications
  • Description:
    Detectable finger joint plasters, blue, refill set for dispenser, 20 plasters
  • L×W:
    - mm
  • Seq:
    05
  • W×D×H:
    160×57×122 mm